Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Cellular ROS/RNS Assay Kit, Cell-Based, 200 Tests

Catalog No. 502583433
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3433 200 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3433 Supplier Abcam Supplier No. AB139473200TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cell-based assay kit for the detection of cellular ROS and RNS in adherent or suspension cells using fluorescence microscopy.

  • Assay type: Cell-based
  • Applications: ICC/IF
  • Sample types: Suspension cells and adherent cells
  • Features: Cited in 11 publications

The product is subject to the following: Abcam Restricted Use Statement

Specifications

For Use With (Equipment) Fluorescence Microscope
Format Kit
Target Molecule Reactive Oxygen Species, Reactive Nitrogen Species
Detection Method Fluorescence
Detection Location In-Cell Detection
No. of Reactions 200 Tests
Sample Type Suspension cells, Adherent cells
Quantity 200 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.