Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cellulase from Aspergillus niger, TCI America™
SDP

Catalog No. C005725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C005725G 25 g
C00571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C005725G Supplier TCI America Supplier No. C005725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Color Very pale yellow - Yellow red at 20°C
Content And Storage 0°C to 10°C
Quantity 25 g
Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.