Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Cellulose Nitrate Membrane Filter
GREENER_CHOICE

Catalog No. 14001955 Shop All Sartorius Products
Change view
Click to view available options
Pore Size:
0.45 μm
0.8 μm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size
14-001-955 0.8 μm
14-001-952 0.45 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-001-955 Supplier Sartorius Supplier No. 11304293N
Only null left
Add to Cart
Add to Cart

Cellulose nitrate membrane filters with 293 mm diameters are hydrophilic, have high flow rates thanks to their symmetrical structure and are compatible with aqueous solutions, hydrocarbons and several other organic solvents.

  • Ideal for several general laboratory applications
  • High, non-specific absorption
  • Hydrophilic, with high flow rates
  • 100 filters
  • 293 mm diameter
TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Filter Type Membrane Filter
Format Disc
Material Cellulose Nitrate
Pore Size 0.8 μm
Sterility Non-sterile
Quantity 100/Cs.
Type Membrane Filter
Diameter (Metric) 293 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.