Learn More
Cytiva (Formerly GE Healthcare Life Sciences) Whatman™ Cellulose Soxhlet Extraction Thimbles, double 2mm wall
Manufactured from high-quality alpha cellulose cotton linter for high-performance use in automated Soxhlet extractor systems. Whatman™ Cellulose Soxhlet Extraction Thimbles are intended for applications including use in air and waste gas analysis.
$203.00 - $351.00
Specifications
Material Cellulose
For Use With (Application) Air and waste gas analysis
View More Specs
Products 7
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Inside Price Quantity & Availability  
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Inside Price Quantity & Availability  
09-657A
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
2810228
80 mm 22 mm
Pack of 25 for $203.00
Add to cart
09-657B
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
2810338
80 mm 33 mm
Pack of 25 for $267.00
Add to cart
09-657H
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
2810432
123 mm 43 mm
Pack of 25 for $351.00
Add to cart
09-657K
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
2810166
60 mm 16 mm
Pack of 25 for $203.00
Add to cart
09-657D
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
2810339
94 mm 33 mm
Pack of 25 for $267.00
Add to cart
09-657P
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
2810266
60 mm 26 mm
Pack of 25 for $216.00
Add to cart
09-657C
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
2810258
80 mm 25 mm
Pack of 25 for $203.00
Add to cart
Description
Description
Manufactured from high-quality alpha cellulose cotton linter for high-performance use in automated Soxhlet extractor systems. Whatman™ Cellulose Soxhlet Extraction Thimbles are intended for applications including use in air and waste gas analysis.
  • Used in a wide variety of solid-liquid extraction studies, including lipid and pesticide analyses in food and soil samples
  • Suitable for temperatures up to 120°C (248°F)
  • Excellent mechanical strength and retention
  • Wall thickness of 2mm to support application need
  • Double thickness for added rigidity
Specifications
Specifications
Cellulose
Air and waste gas analysis