Learn More
Description
CellVation is a cryopreservation medium designed for optimal cell recovery and convenience. It does not contain DMSO or serum, and is scientifically engineered for serum-free cultures, as well as, ideally suited for all types of serum. It has also been shown to preserve the potential of stem cells for differentiation.
Specifications
Specifications
| Form | Solution |
| Product Type | Cryopreservation Media |
| For Use With (Application) | Cell counting, viability and cryopreservation |
| Quantity | 1 L |
| Certifications/Compliance | Mycoplasma, Bacteria, Fungi, Yeast and Mold free. Certified U.S. origin. |
| Endotoxin Level | <0.5 EU/mL |
| pH | 6.5 to 7.5 |
| Content And Storage | Store at 4°C |
| Description | CellVation Cryopreservation Media |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.