Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One CELLview™ Sterile Cell Culture Dishes With Glass Bottom

Catalog No. 07000236 Shop All Greiner Bio-One Products
Change view
Click to view available options
Surface Treatment:
Advanced TC
Non-treated
Tissue Culture
Well Count:
1
4
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Surface Treatment Well Count
07-000-236 Non-treated 4
07-000-235 Tissue Culture 4
07-000-583 Advanced TC 1
07-000-582 Tissue Culture 1
07-000-234 Advanced TC 4
07-000-233 Non-treated 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 07-000-236 Supplier Greiner Bio-One Supplier No. 627871
This item is not returnable. View return policy

Free of detectable DNase, RNase, human DNA. Greiner Bio-One CELLview™ Cell Culture Dishes With Glass Bottom features high transparent achromatic borosilicate glass; hydrolytic class 1 (DIN-ISO 719).

  • Non-pyrogenic
  • Non-cytotoxic
  • Free of detectable DNase, RNase, human DNA
  • High transparent achromatic borosilicate glass
  • Maximal spectral transmission, no autofluorescence, no depolarisation of light
  • Available

Certifications

ISO 8255-1:1986

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1.5 mL/Compartment
Culture Area 1.9 sq. cm/compartment
Material Polystyrene/Glass Bottom
Sterility Sterile
Surface Treatment Non-treated
Well Count 4
Dnase Free Yes
RNase Free Yes
Thickness Glass bottom thickness: 175 µm +/- 15 µm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.