Learn More
Invitrogen™ CellVue™ Burgundy Cell Labeling Kit
Catalog No. 5017153
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
Catalog No. Quantity
50-171-53 1 kit
1 options
Catalog No. 50-171-53 Supplier Invitrogen™ Supplier No. 88087216
  • $387.00
    $290.65 / Each of 1
    Save $96.35
    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart
Far-red/near-infrared fluorescent reagent used to label the cell membrane in order to identify and track labeled cells. Cell labeling can be combined with fluorescently labeled antibodies and other markers of cellular function for flow cytometric analysis and fluorescent microscopy.
  • The Mini CellVue™ Kits are supplied with one vial of dye stock (1 mM in ethanol) and one vial of labeling vehicle (Diluent C)
  • CellVue™ Burgundy is optimally excited at 683nm and has a peak emission of 707nm
  • It can be detected equally in filter sets designed for Alexa Fluor™ 700 and APC-eFluor™ 780 or similar fluorochromes – using antibodies conjugated to these fluorochromes in combination with CellVue™ Burgundy is not recommended
  • Burgundy can be used in combination with APC
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 kit
Reagent Type Cell Tracker Compounds, Cell Labeling Reagents
Label Type eFluor
Product Type Cell Labeling
For Research Use Only. Not for use in diagnostic procedures.