Learn More
Invitrogen™ CellVue™ Lavender Cell Labeling Kit
Catalog No. 5017155
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
Catalog No. Quantity
50-171-55 1 kit
1 options
Catalog No. 50-171-55 Supplier Invitrogen™ Supplier No. 88087316
Add to Cart
Edge
Add to Cart
Violet fluorescent reagent used to label the cell membrane in order to identify and track labeled cells. Cell labeling can be combined with fluorescently labeled antibodies and other markers of cellular function for flow cytometric analysis and fluorescent microscopy.
  • The Mini CellVue™ Kits are supplied with one vial of dye stock (1 mM in ethanol) and one vial of labeling vehicle (Diluent C)
  • CellVue™ Lavender is optimally excited at 420nm and has a peak emission of 461nm
  • CellVue™ Lavender is compatible with most multi-color flow cytometric applications; however, due to similar emission properties it cannot be used in combination with eFluor™ 450 or Pacific Blue™ reagents
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 kit
Reagent Type Cell Tracker Compounds, Cell Labeling Reagents
Product Type Cell Labeling
For Research Use Only. Not for use in diagnostic procedures.