Learn More
Invitrogen™ CellVue™ NIR815 Cell Labeling Kit
Catalog No. 5017157
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
Catalog No. Quantity
50-171-57 1 kit
1 options
Catalog No. 50-171-57 Supplier Invitrogen™ Supplier No. 88087416
Add to Cart
Add to Cart
Near-infrared fluorescent reagent used to label the cell membrane in order to identify and track labeled cells. Cell labeling can be combined with fluorescently labeled antibodies and other markers of cellular function for flow cytometric analysis and fluorescent microscopy.
  • The Mini CellVue™ Kits are supplied with one vial of dye stock (1 mM in ethanol) and one vial of labeling vehicle (Diluent C)
  • CellVue™ NIR815 is optimally excited at 786nm and has a peak emission of 814nm
  • CellVue™ NIR815 has been reported to be useful for short-term in vivo cell tracking studies using a Pearl™ Imager or Odyssey Infrared Imaging System (Li-COR Biosciences, Inc., Lincoln, NE)
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 kit
Reagent Type Cell Tracker Compounds, Cell Labeling Reagents
Product Type Cell Labeling
For Research Use Only. Not for use in diagnostic procedures.