Learn More
Invitrogen™ CellVue™ Plum Cell Labeling Kit
Catalog No. 5017151
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
Catalog No. Quantity
50-171-51 1 kit
1 options
Catalog No. 50-171-51 Supplier Invitrogen™ Supplier No. 88087116
Add to Cart
Edge
Add to Cart
Far-red fluorescent reagent used to label the cell membrane in order to identify and track labeled cells. Cell labeling is rapid and stable and can be combined with fluorescently labeled antibodies and other markers of cellular function for flow cytometric analysis and fluorescent microscopy.
  • The Mini CellVue™ Kits are supplied with one vial of dye stock (1 mM in ethanol) and one vial of labeling vehicle (Diluent C)
  • CellVue™ Plum is optimally excited at 652nm and has a peak emission of 671nm
  • It can be detected equally in filter sets designed for APC, Alexa Fluor™ 700, and APC-eFluor™ 780 or similar fluorochromes – using antibodies conjugated to these fluorochromes in combination with CellVue™ Plum is not recommended
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 kit
Reagent Type Cell Tracker Compounds, Cell Labeling Reagents
Label Type eFluor
Product Type Cell Labeling
For Research Use Only. Not for use in diagnostic procedures.