Learn More
R&D Systems™ CellXVivo Human Monocyte-derived DC Differentiation Kit
GSA_VASDP
Catalog No. CDK004 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
CDK004 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. CDK004 Supplier R&D Systems™ Supplier No. CDK004
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
Includes
Serum-Free Dendritic Cell Base Media; Recombinant Human IL-4; Recombinant Human GM-CSF; Recombinant Human TNF-alpha; Reconstitution Buffer 2
For the differentiation of monocyte-derived dendritic cells
For the differentiation of CD14+ monocytes into dendritic cells.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Line R&D Systems
Quantity 1 Kit
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.