Learn More
Bullard™ CEN10™ Sleek Low Profile Helmet
SDP
Catalog No. 19160569 Shop All Bullard Company Products
Change view
Click to view available options
Ventilated:
Not Vented
Vented
Color:
Dove Gray
Hi-vis Yellow
Orange
Red
White
Yellow
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Ventilated Color
19-160-569 Vented Dove Gray
19-160-570 Vented Yellow
19-160-560 Not Vented White
19-160-561 Not Vented Red
19-160-562 Not Vented Dove Gray
19-160-563 Not Vented Yellow
19-160-564 Not Vented Hi-vis Yellow
19-160-565 Not Vented Orange
19-160-567 Vented White
19-160-568 Vented Red
19-160-571 Vented Hi-vis Yellow
19-160-572 Vented Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 19-160-569 Supplier Bullard™ Supplier No. C10VDGAMR
Add to Cart
Add to Cart
Bullard CEN10™ Sleek Low Profile Helmet features sleek, low-profile vented and non-vented designs, self-adjusting patented FlexGlide™ suspension system, an impact resistant helmet shell with inner foam comfort liner, and an integrated 3-point chinstrap.
  • The patented FlexGlide suspension system lets wearers auto-adjust the fit of their helmet for all day comfort
    • The CEN10 is the only auto-adjusting suspension helmet on the market, allowing even the smallest adjustments to achieve the perfect fit
  • Replaceable brow pad, comfort liner, and nape cover for ultimate utility
  • Distinctive three rib trademark
  • Manufacturer warranty: 3 years
  • Life span: 5 years
Specifications
Certifications/Compliance ANSI/ISEA Z87.1
Color Dove Gray
Dimensions (L x W x H) 11 x 8.75 x 6.25 in.
Liner Material EPS Foam
Material PC ABS Blend
Size Adjustable
Ventilated Vented
Head Style FlexGuide Head Suspension System
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.