Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CENPA Monoclonal Antibody (3-19)
GREENER_CHOICE

Catalog No. PIMA120832 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA120832 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA120832 Supplier Invitrogen™ Supplier No. MA120832
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Recommended positive controls: , BE cells. This antibody does not cross react with mouse CENPA proteins. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Centromeres are the differentiated chromosomal domains that specify the mitotic behavior of chromosomes. CENPA encodes a centromere protein which contains a histone H3 related histone fold domain that is required for targeting to the centromere. CENPA is proposed to be a component of a modified nucleosome or nucleosome-like structure in which it replaces 1 or both copies of conventional histone H3 in the (H3-H4)2 tetrameric core of the nucleosome particle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CENPA
Applications ChIP Assay, Immunohistochemistry (Paraffin), Neutralization, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3-19
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and no preservative; pH 7.2
Gene Cenpa
Gene Accession No. P49450, Q6XXM1
Gene Alias CenH3; CENP A; CENPA; CENP-A; Centromere autoantigen A; centromere protein A; centromere protein A, 17kDa; centromere-specific histone; ggCENP-A; histone H3-like centromeric protein A; RGD1563607
Gene Symbols Cenpa
Host Species Mouse
Immunogen Synthetic peptide: PRRRSRKPEAPRRRSPS, corresponding to amino acids 3-19 of Human CENP A.
Purification Method Protein A
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100113359, 1058
Target Species Human, Chicken, C. elegans
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.