Learn More
Invitrogen™ CENPB Monoclonal Antibody (A5F6)

Description
Positive Control: Hela cell lysate, A549 cell lysate, MCF-7 cell lysate, SH-SY5Y, human kidney tissue, human skin tissue. Subcellular Location: Nucleus, centromere.
Specifications
Specifications
| Antigen | CENPB |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | A5F6 |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | CENPB |
| Gene Accession No. | P07199 |
| Gene Alias | CENPB; CENP-B; CENP-B protein; centromere autoantigen B; centromere protein B; centromere protein B, 80kDa; major centromere autoantigen B |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.