Learn More
Invitrogen™ CENPF Polyclonal Antibody

Description
Suggested positive control: HeLa and SK-N-SH cells.
Specifications
Specifications
| Antigen | CENPF |
| Applications | Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene | CENPF |
| Gene Accession No. | P49454 |
| Gene Alias | 6530404A22Rik; AH antigen; AI325968; cell-cycle-dependent 350K nuclear protein; CENF; CENPF; CENP-F; CENP-F kinetochore protein; centromere autoantigen F; centromere protein F; centromere protein F, (mitosin); centromere protein F, 350/400ka (mitosin); centromere protein F, 350/400kDa (mitosin); CILD31; hcp-1; im:7140452; im:7144238; im:7154719; im:7156761; kinetochore protein CENPF; Lek1; leucine, glutamic acid, lysine family 1 protein; mitosin; PRO1779; RP11-262H5.1; si:ch211-152h14.2; STROMS |
| Gene Symbols | CENPF |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.