Learn More
CELLTREAT Scientific Products 4mL Centrifugal Filter Unit, MWCO 50kDa, Non-Sterile
SDP
Supplier:  CELLTREAT Scientific Products 230714
Encompass_Preferred
Filter cartridges feature PES membranes with 5, 10, 30, 50 or 100kDa size exclusion cut-offs Large active membrane area with dual vertical design for fast and efficient processing Centrifuge tubes feature internal supports to securely position the filter cartridge Applications include sample concentration, molecule purification, protein fractionation, buffer exchange, and desalting Cartridge design prevents accidental over-concentration of samples High sample recovery of >90% Graduations on conical tube and filter cartridge are printed in bold, easy-to-read black ink Large white marking area provides excellent contrast for identification Filter cartridges are clearly printed with Molecular Weight Cut Off (MWCO) in daltons Easy to write on flat surface HDPE caps feature integrated polyethylene liners for a leak-proof seal Molded graduations in the conical bottom are ideal for small volume quantification Standard centrifuge tube rotor compatibility
Catalog No. NC2544546
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.