Learn More
Foxx Life Sciences EZBio Centrifuge Tube Assemblies, 50mL, Sterile, Single Use Technology with EZBio Tubing and MPC CPC, Sterile, 10/CS
Supplier: Foxx Life Sciences 5853001FLS
Introducing the EZBio Centrifuge Tube Assembly from Foxx Life Sciences—a closed-system solution with TPE tubing and a vent filter. Designed for biopharmaceutical sampling and transfer, it comes in 15mL and 50mL volume.
Molded from USP Class VI Resin and Gamma Irradiated in the U.S. for sterility, the assembly includes a polypropylene centrifuge tube, an 18” inlet line with pinch clamp and plug, a 1.5” vent line with PTFE hydrophobic filter, diptube and stand for secure holding.
Features
Closed-system design ensures contamination-free handling.
Leak-proof seal with platinum-cured silicone gasket.
Molded from USP Class VI Resin, FDA and USP-compliant polypropylene.
Available in 15mL and 50mL, with 6 TPE tubing options and 4 fittings.
Applications
Cell Harvest and Downstream Processing Intermediates
Drug Substance Purification
Drug Product Formulation
Buffer Preparation
Media Preparation
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.