Learn More
Invitrogen™ Centaurin alpha-2 Polyclonal Antibody

Description
Immunogen sequence: MGDRERNKKR LLELLRAPDT GNAHCADCGA ADPDWASYKL GIFICLNCCG VHRNFPDISR VKSVRLDFWD DSIVEFMIHN GNLRVKAKFE ARVPAFYYIP QANDCLVLKE QWIRAKYERR EFMADGETIS LPGNREGFLW KRGRDNSQFL RRKFVLLARE GLLKYFTKEQ GKSPKAVISI KDLNATFQTE KIGHPHGLQI TYRRDGHTRN LFVYHESGKE IVDWFNALRA ARLQYLKMAF PELPESELVP FLTRNYLKQG FMEKTGPKQK EPFKKRWFAL DCHERRLLYY KNPLDAFEQG QVFLGNKEQG YEAYEDLPKG IRGNRWKAGL TIVTPERRFV LTCPSEKEQQ EWLESLRGVL SSPLTPLNRL TASTESGRSS R; Positive Samples: Mouse skeletal muscle; Cellular Location: Cell membrane, Cytoplasm.
Specifications
Specifications
| Antigen | Centaurin alpha-2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.37 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | ADAP2 |
| Gene Accession No. | Q8R2V5 |
| Gene Alias | Adap2; Arf-GAP with dual PH domain-containing protein 2; ArfGAP with dual PH domains 2; CENTA2; centaurin; centaurin beta; centaurin, alpha 2; centaurin-alpha 2 protein; centaurin-alpha-2; Centaurin-alpha2 protein; cent-b; cnt-a2; HSA272195 |
| Gene Symbols | ADAP2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.