Learn More
OHAUS Centrifuge Micro
GSA_VA
Catalog No. 01920171 Shop All OHAUS Products
Change view
Click to view available options
Capacity:
24 x 1.5/2 mL
Catalog No. Capacity
01-920-171 24 x 1.5/2 mL
1 options
Catalog No. 01-920-171 Supplier OHAUS Supplier No. 83042060
Add to Cart
Add to Cart
Request a Quote
Frontier™ micro centrifuges deliver high-speed operation for a range of essential lab applications, such as DNA/RNA preparation, ultrafiltration, and more. Biocontainment rotors enable research involving hazardous samples.
  • Excellent functionality when used with rotors compatible with 1.5/2.0 mL or 5 mL microtubes with high throughput and biocontainment options, spin columns, PCR strips, cryo tubes and hematocrit capillaries.
  • Designed for ease of operation and feature convenient controls on an intuitive interface, allowing responsive operation, even when wearing gloves.
  • Packaging all their functionality within a small footprint, ;these centrifuges save valuable space on the lab bench.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Capacity 24 x 1.5/2 mL
Max. Speed 14,500 rpm
Max. RCF 17,135 x g
Refrigerated Yes
Product Type Centrifuge