Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Centrifuge Tube
SDP

Catalog No. STLLX4115
Change view
Click to view available options
Capacity (Metric):
15 mL
50 mL
Sterility:
Non-sterile
Sterile
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Sterility
STLLX4115 15 mL Non-sterile
STLLX4103 50 mL Non-sterile
STLLX4108 50 mL Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. STLLX4115 Supplier StatLab Supplier No. LX4115
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Cytology centrifuge tubes.

  • Graduated
  • Conical bottom
  • Translucent polypropylene, packed in styrofoam racks
  • Screw-on caps provide firm, leak-resistant fit

Specifications

Capacity (Metric) 15 mL
Disposable Single-use
Material Polypropylene
Includes 15 mL Sterile Centrifuge Tubes
Sterility Non-sterile
Autoclavable Not Autoclavable
Shape Cylindrical
Bottom Shape Conical
Closure Type Screw Cap
Closure Included Yes
Writing Area Yes
Graduated Yes
Product Type Clinical Centrifuge Tube
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.