Learn More
Electron Microscopy Sciences Flat Screw Cap Conical Centrifuge Tube, 15 ml, Foam Rack in Sterile Bag, 17 x 118 mm
Supplier: Electron Microscopy Sciences 6302601
Centrifuge tubes made of premium polypropylene for clarity and centrifugal strength. They are supplied sterile, and although they are autoclavable, their polyethylene screw caps are not. Tubes and caps may be frozen at temperatures as low as -45 deg C. Caps have a flat top surface for easy marking. Sterile, Rnase, Dnase, pyrogen, and cytotoxin free. Compatible with standard rotors. Frosted marking surface. Maximum centrifugal force is 12,000xg.
Volume: 15 ml
Package: Foam Rack in Sterile Bag
Size: 17 x 118 mm
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.