Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Centrin 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140246
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140246 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140246 Supplier Invitrogen™ Supplier No. PA5140246
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: ASGFKKPSAA STGQKRKVAP KPELTED Highest antigen sequence indentity to the following orthologs: Rat - 67%, Mouse - 70%.

The protein encoded by this gene plays important roles in the determination of centrosome position and segregation, and in the process of microtubule severing. This protein is localized to the centrosome of interphase cells, and redistributes to the region of the spindle poles during mitosis, reflecting the dynamic behavior of the centrosome during the cell cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Centrin 1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CETN1
Gene Accession No. Q12798
Gene Alias calcium binding protein; Calt; Caltractin; Caltractin isoform 2; CEN1; centrin 1; centrin 1 S homeolog; centrin, EF-hand protein, 1; centrin, EF-hand protein, 2; centrin-1; CETN; cetn1; cetn1.S; cetn2; EF-hand protein; testicular tissue protein Li 37; testis-specific murine centrin; Unknown (protein for MGC:133570); xcen; XELAEV_18042289mg
Gene Symbols CETN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CETN1. Recombinant protein control fragment (Product #RP-109178).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1068
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.