Description
Includes
Instructions
- The weight of one to 20 plated washers hanging from a nylon thread is balanced against the pull on the other end of the thread by a rubber stopper swung in a horizontal circle
- The thread runs with minimum friction through a polished plastic tube
Specifications
Specifications
| Product Type | Kit, Centripetal Force Apparatus |
| For Use With (Application) | Measuring centripetal force |
| Includes | Low-friction guide, stout cord, resilient mass and washer masses and Instruction |