Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Centripetal Force Kit
SDPSCI_ED

Catalog No. S89542 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S89542 5 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy

Prop 65 Warning: Cancer and reproductive harm

Catalog No. S89542 Supplier United Scientific™ Supplier No. UNCFKIT02
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Prop 65 Warning: Cancer and reproductive harm

Help students to investigate the force on an object in circular motion with United Scientific™ Centripetal Force Kit. An activity guide is included.

Help students to investigate the force on an object in circular motion with United Scientific Centripetal Force Kit. Through a series of simple demonstrations, students will better understand the relationship between the radius of the circular motion, the velocity of the moving object, and the external force required to maintain the motion. Contents:

  • Rubber stopper attached to cord approximately 1 m in length
  • Plastic tube with beveled ends
  • Hanging mass set: 12 aluminum weights - 9 x 20 g, 1 x 10 g, 2 x 5 g
  • One 50 g aluminum hanger
  • Paperclip (position marker)
  • Activity guide included

Specifications

Grade 5 to 12
Format Kit
Product Type Centripital Force Kit
Languages English
Subject Physics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.