Learn More
Centrisart™ I centrifugal ultrafiltration unit, Sold by MilliporeSigma™ Supelco™
Catalog No. 111021602 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Pore Size:
100 kDa MWCO
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size
11-102-1602 -
11-102-1604 100 kDa MWCO
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-1602 Supplier MilliporeSigma Supelco Supplier No. 13239E
Add to Cart
Add to Cart
Their unique design prevents membrane blockage when doing protein-free ultrafiltration.
TRUSTED_SUSTAINABILITY
Specifications
Pore Volume 2.5 mL
Quantity 12 Tubes
Surface Area 0.79 cm2
Volume (Metric) 15 mL
Material Polystyrene
Centrisart is a registered trademark of Sartorius Stedim Biotech GmbH