Learn More
Labconco CentriVap™ Cart
GREENER_CHOICESDP
Catalog No. 12199549 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Labconco CentriVap Pro and Standard Vacuum Concentrator Systems
Catalog No. For Use With (Equipment)
12-199-549 Labconco CentriVap Pro and Standard Vacuum Concentrator Systems
1 options
Catalog No. 12-199-549 Supplier Labconco Supplier No. 8544000
Add to Cart
Add to Cart
Powder-coated steel cart supports CentriVap™ Vacuum Concentrators, CentriVap™ Cold Traps, and vacuum pumps during use, storage, and transportation.
  • Ergonomic design allows for easy access to all key components of the system.
  • Shipping weight 132.0 lb. (59.9 kg)
Specifications
Material Stainless Steel
Type Rotor
For Use With (Equipment) Labconco CentriVap Pro and Standard Vacuum Concentrator Systems