Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Labconco CentriVap™ Cold Trap with Acrylic Lid

Designed to collect evaporated liquid from a CentriVap™ Concentrator, protecting the vacuum pump and lab environment from vapors and fumes.
$6165.00
Catalog Number | Mfr. No. | Voltage | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Voltage | Price | Quantity & Availability | |||||
12-199-527
![]() |
Labconco
8601010 |
230 V |
Each for $6,165.00
|
|
|||||
12-199-514
![]() |
Labconco
8601000 |
115 V |
Each for $6,165.00
|
|
|||||
Description
- Brushed, stainless steel and powder-coated steel exterior
- Illuminated temperature graph with endpoint indication
- HCFC-free refrigerant system
- 4 L stainless steel collector
- Acrylic lid with 2 quick-disconnect fittings for connection to a CentriVap™ concentrator
- Stainless steel drain port (with corrosion-resistant, nylon valve)
- -50 °C models feature a 1/3 hp compressor (-50 °C in less than 30 min.)
Specifications
Stainless Steel | |
Cold Trap | |
North American |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title