Learn More
Labconco CentriZap Pro Strobe Light
GREENER_CHOICESDP
Catalog No. 12199519 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Labconco CentriVap Pro and Standard Vacuum Concentrator Systems
Catalog No. For Use With (Equipment)
12-199-519 Labconco CentriVap Pro and Standard Vacuum Concentrator Systems
1 options
Catalog No. 12-199-519 Supplier Labconco Supplier No. 8547200
Add to Cart
Add to Cart
Enables users to view samples as if they were stationary while rotor is spinning, eliminating the need to pause a cycle.
  • Includes mounting holster and hardware for attachment to the right side of the CentriVap™ Pro Vacuum Concentrator.
  • Shipping weight 2.0 lb. (0.9 kg)
Specifications
Type Strobe Light
For Use With (Equipment) Labconco CentriVap Pro and Standard Vacuum Concentrator Systems