Description
- Wide-angle wraparound lens provides indirect ventilation for chemical splash protection
- Allows excellent peripheral vision and impact resistance
- Fits comfortably over most prescription glasses
- 3M Catalog# 40304-00000-10 , 3M Catalog# 40305-00000-10
Order Info
Available with an optional antifog lens.
Compliance
Meet ANSI Z87.1-2010 and CSA Z94.3 standards
Specifications
Specifications
| Lens Coating | Impact-resistant |
| Lens Type | Chemical-resistant |
| Certifications/Compliance | ANSI Z87.1-2010 and CSA Z94.3 standards |
| Lens Tint | Clear |
| Product Type | Safety goggles |
| Ventilation Details | Indirect |