Learn More
CEP First Responder Spill Kits
SDP
Catalog No. 191203616
Click to view available options
Size:
2 Responders
4 Responders
6 Responders
8 Responders
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-120-3616 6 Responders
19-120-3615 4 Responders
19-120-3614 2 Responders
19-120-3617 8 Responders
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-120-3616 Supplier CEP Supplier No. CEPHAZRESPONDER6
Add to Cart
Add to Cart
Request a Quote
Be ready for fast, safe response to chemical emergencies with these universal response kits
Includes all necessary supplies to safely contain and clean up oils, acids, caustics, solvents, and toxic spills; also includes protective apparel for the number of persons indicated in the kit size (2, 4, 6 or 8 responders).
  • One 95-gal. poly overpack drum
  • 100 17 × 19 in. universal pads
  • Four 5 × 10 in. universal booms
  • Four epoxy sticks
  • One pipe wrap kit
  • Two hazardous waste labels
  • One nonsparking shovel
  • One 36 × 36 in. polyurethane drain seal
  • One quart each of acid and caustic neutralizer
  • Coveralls with hood and booties, nitrile gloves, safety goggles, and disposal bags to accommodate number of persons designated in kit size
Specifications
Capacity (English) 95 gal.
Capacity (Metric) 359.6 L
For Use With (Application) Clean up oils, acids, caustics, solvents, and toxic spills
Includes 95 gal. poly overpack drum, (4) 5 x 10 in. universal booms, 4 epoxy sticks, 1 pipe wrap kit, 2 hazardous waste labels, 1 non-sparking shovel, 36 x 36 in. polyurethane drain seal, 1 quart each of acid and caustic neutralizer
Size 6 Responders