Learn More
CEP Floor Sorbents
SDP
Catalog No. 191301980
Click to view available options
Type:
Cellulose Sorbent
Vermiculate Sorbent
Weight (English):
20 lb.
22 lb.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Weight (English)
19-130-1980 Vermiculate Sorbent 22 lb.
19-130-3847 Cellulose Sorbent 20 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-130-1980 Supplier CEP Supplier No. CEPVERM4
Add to Cart
Add to Cart
Specifications
Type Vermiculate Sorbent
For Use With (Application) Solidification, packaging, incineration
Weight (English) 22 lb.
Weight (Metric) 10 kg