Learn More
CEP131/AZ1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301416A
Description
The recommended shelf life for this product is 1 year from date of receipt.
CEP131 (Centrosomal Protein 131) is a Protein Coding gene. Diseases associated with CEP131 includeAnterior Spinal Artery Syndrome. Among its related pathways areOrganelle biogenesis and mainteceandCell Cycle, Mitotic. Gene Ontology (GO) annotations related to this gene includeprotein homodimerization activity.Specifications
| CEP131/AZ1 | |
| Polyclonal | |
| Unconjugated | |
| CEP131 | |
| 5-azacytidine induced 1, 5-azacytidine-induced protein 1, AZ1, AZI1, centrosomal protein 131 kDa, centrosomal protein 131kDa, KIAA1118, pre-acrosome localization protein 1, ZA1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22994 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q9UPN4 | |
| CEP131 | |
| Between 325 and 375 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.