Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEP164 Monoclonal Antibody (17)
GREENER_CHOICE

Catalog No. PIMA531584
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIMA531584 100 μL
1 options

Catalog No. PIMA531584

Supplier: Invitrogen™ MA531584

Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive Control: HeLa, A549.

CEP164 was initially identified as a centrosomal protein, but other studies have indicated that it also plays a role in the formation of primary cilia, the microtubule-based sensory antennae projecting from the surface of many eukaryotic cells as well as in DNA damage response acting as a mediator protein. CEP164 interacts with both ATR and ATM, proteins that trigger a number of cellular responses including the initiation of DNA damaged-induced cell cycle checkpoints. It is phosphorylated upon replication stress, ultraviolet (UV) radiation, and ionizing radiation silencing of CEP164 significantly reduces the DNA damage-induced phosphorylation of several proteins in the DNA damage-activated signaling cascade and compromises cell survival after UV damage. At least two isoforms of CEP164 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEP164
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 17
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CEP164
Gene Accession No. Q9UPV0
Gene Alias AI450905; BC027092; ce164; centrosomal protein 164; centrosomal protein 164kDa; centrosomal protein of 164 kDa; Cep 164; CEP164; D030051D21; KIAA1052; mKIAA1052; NPHP15; RGD1560988; RGD1561243
Gene Symbols CEP164
Host Species Mouse
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22897
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.