Learn More
CEP170 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For ICC, formaldehyde fixation is recommended. Permeabilization with Triton-X 100 is recommended for formaldehyde-fixed cells.
Specifications
Specifications
| Antigen | CEP170 |
| Applications | ChIP sequencing (ChIP-seq), Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/ml |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.1% BSA and 0.09% sodium azide |
| Gene | CEP170 |
| Gene Accession No. | Q5SW79 |
| Gene Alias | Centrosomal protein of 170 kDa, Cep170, KARP-1-binding protein, KARP1-binding protein, CEP170, FAM68A, KAB, KIAA0470 |
| Gene Symbols | CEP170 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.