Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEP170L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549088
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549088 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549088 Supplier Thermo Scientific Supplier No. PA549088
Only null left
Add to Cart
Add to Cart

CEP170L Polyclonal Antibody

CEP170L Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen CEP170L
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CEP170P1
Gene Accession No. Q96L14
Gene Alias centrosomal protein 170 pseudogene 1; CEP170 pseudogene 1; CEP170L; Cep170-like protein; CEP170P1; FAM68B; KIAA0470L
Gene Symbols CEP170P1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 140-167 amino acids from the Central region of human CEP170L.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 645455
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.