Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEP192 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABE1965MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE1965MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1965MI Supplier MilliporeSigma Supplier No. ABE1965

Rabbit Polyclonal Antibody

Anti-CEP192 is a highly specific rabbit polyclonal antibody that targets Centrosomal protein of 192 kDa and has been tested in Flow Cytometry and Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEP192
Applications Flow Cytometry, Immunofluorescence
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Cep192,KIAA1569,PP8407
Host Species Rabbit
Immunogen His-tagged recombinant fragment corresponding to 299 amino acids from the C-terminal region of human Centrosomal protein of 192 kDa (CEP192).
Purification Method Affinity Purified
Quantity 200 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_115518
Test Specificity This polyclonal antibody detects CEP192 in human. It targets an epitope within 299 amino acids from the C-terminal region.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.