Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEP250/C-NAP1 Mouse anti-Human, Clone: 2G12D12, Proteintech
SDP

Catalog No. 501737205 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7205 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7205 Supplier Proteintech Group Inc Supplier No. 668141IG150UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Probably plays an important role in centrosome cohesion during interphase.

Specifications

Antigen CEP250/C-NAP1
Applications Western Blot
Classification Monoclonal
Clone 2G12D12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CEP250
Gene Accession No. Q9BV73
Gene Alias 250 kDa centrosomal protein, C NAP1, Centrosomal protein 2, centrosomal protein 250kDa, CEP2, CEP250, CNAP1
Gene Symbols CEP250
Host Species Mouse
Immunogen CEP250,C-NAP1 Fusion Protein Ag5925
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11190
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.