Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEP290 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501560931
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-0931 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-0931 Supplier Bethyl Laboratories, Inc Supplier No. A301659AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For ICC, formaldehyde fixation is recommended. Permeabilization with Triton-X 100 is recommended for formaldehyde-fixed cells.

CEP290 activates ATF4 mediated transcription and is required for the correct localization of ciliary and phototransduction proteins in retinal photoreceptor cells; may play a role in ciliary transport processes. CEP290 is ubiquitously expressed; strongly in placenta and weakly in brain. There are two named isoforms. Defects in CEP290 are a cause of Joubert syndrome type 5 (JBTS5), Senior-Loken syndrome type 6 (SLSN6), Leber congenital amaurosis type 10 (LCA10) and Meckel syndrome type 4 (MKS4). Antibodies against CEP290 are present in sera from patients with cutaneous T cell lymphomas, but not in the healthy control population.

Specifications

Antigen CEP290
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene CEP290
Gene Accession No. O15078
Gene Alias Centrosomal protein of 290 kDa, Cep290, Bardet-Biedl syndrome 14 protein, Cancer/testis antigen 87, CT87, Nephrocystin-6, Tumor antigen se2-2, CEP290, BBS14, KIAA0373, NPHP6
Gene Symbols CEP290
Host Species Rabbit
Immunogen Between 2429 and 2479
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80184
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.