Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEP350 Monoclonal Antibody (CL3423)
GREENER_CHOICE

Catalog No. PIMA531385
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIMA531385 100 μL
1 options

Catalog No. PIMA531385

Supplier: Invitrogen™ MA531385

Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: QVVQSQREVT EVLQEATCKI AAQQSETARL TTDAARQICE MAELTRTHIS DAVVASGAPL AILYDHQRQH LPDFVKQLRT RTETDRKSPS VSLSQ Highest antigen sequence indentity to the following orthologs: Mouse - 81%, Rat - 83%.

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEP350
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Monoclonal
Clone CL3423
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CEP350
Gene Accession No. Q5VT06
Gene Alias 4933409L06Rik; 6430546F08Rik; Cap350; centrosomal protein 350; centrosomal protein 350kDa; centrosome-associated protein 350; Centrosome-associated protein of 350 kDa; CEP350; GM133; Kiaa0480
Gene Symbols CEP350
Host Species Mouse
Immunogen Recombinant protein fragment of Human CEP350 (Product #RP-105842)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9857
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.