Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ CEP350 Recombinant Protein Antigen
SDP

Catalog No. NBP255557PE Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP255557PE 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP255557PE Supplier Novus Biologicals™ Supplier No. NBP255557PEP

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human CEP350. Source: E.coli Amino Acid Sequence: QVVQSQREVTEVLQEATCKIAAQQSETARLTTDAARQICEMAELTRTHISDAVVASGAPLAILYDHQRQHLPDFVKQLRTRTETDRKSPSVSLSQ The CEP350 Recombinant Protein Antigen is derived from E. coli. The CEP350 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.

Specifications

Gene ID (Entrez) 9857
Purification Method >80% by SDS-PAGE and Coomassie blue staining
Common Name CEP350 Recombinant Protein Antigen
Content And Storage Store at −20°C. Avoid freeze-thaw cycles.
Formulation PBS and 1M Urea, pH 7.4.
For Use With (Application) Blocking/Neutralizing, Control
Gene Alias CAP350GM133, centrosomal protein 350kDa, centrosome-associated protein 350, Centrosome-associated protein of 350 kDa, Cep350, FLJ38282, FLJ44058, KIAA0480centrosome associated protein 350
Gene Symbol CEP350
Label Type Unlabeled
Product Type Recombinant Protein Antigen
Quantity 100 μL
Regulatory Status RUO
Source E.Coli
Specific Reactivity This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-51200. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.