Learn More
Invitrogen™ CEP41 Polyclonal Antibody

Description
Immunogen sequence: HIVGAYSYPI ATLSRTMNPY SNDILEYKNA HGKIIILYDD DERLASQAAT TMCERGFENL FMLSGGLKVL AQKFPEGLIT GSLP Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 99%.
Specifications
Specifications
| Antigen | CEP41 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CEP41 |
| Gene Accession No. | Q9BYV8 |
| Gene Alias | 1700017E11Rik; 2810431D15Rik; centrosomal protein 41; centrosomal protein 41 kDa; centrosomal protein 41kDa; centrosomal protein of 41 kDa; CEP41; JBTS15; testis specific gene A14; testis specific protein A14; testis specific, 14; testis-specific gene A14 protein; Tsga14 |
| Gene Symbols | CEP41 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.