Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cephalexin Monohydrate USP MP Biomedicals

Catalog No. ICN15058510 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15058510 10 g
ICN15058505 5 g
ICN15058525 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15058510 Supplier MP Biomedicals, Inc Supplier No. 0215058510
Add to Cart
Add to Cart

Chemical Identifiers

CAS 23325-78-2
Molecular Formula C16H19N3O5S
Molecular Weight (g/mol) 365.40
MDL Number MFCD00167148,MFCD00056877
InChI Key AVGYWQBCYZHHPN-CYJZLJNKSA-N
Synonym 7-(d-a-Aminophenylacetamido)desacetoxycephalosporanic acid, Cefalexin monohydrate
PubChem CID 62921
ChEBI CHEBI:3535
IUPAC Name (6R,7R)-7-[(2R)-2-amino-2-phenylacetamido]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrate
SMILES O.CC1=C(N2[C@H](SC1)[C@H](NC(=O)[C@H](N)C1=CC=CC=C1)C2=O)C(O)=O

Specifications

Type Cephalexin Monohydrate
Physical Form Crystalline Powder
Color Beige to White
Quantity 10 g
Formula Weight 365.4
Grade BP/USP
Assay ≥95%
Chemical Name or Material Cephalexin Monohydrate
EINECSNumber 239-773-6
Recommended Storage Store at 4°C, desiccate.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.