Learn More
AdipoGen Cephalochromin
SDP
Supplier:  AdipoGen BVT0440M005
Encompass_Preferred
Chemical. CAS 25908-26-3. Formula C28H22O10. MW 518.5. Isolated from Pseudoanguillospora sp. strain 6577. Mycotoxin. Bisnaphtha-gamma-pyrone derivative similar to chaetochromin and ustilaginoidin. Antibiotic. Antibacterial, antifungal and anti-algal compound. Antitumor and cytotoxic compound. Calmodulin-sensitive PDE activity inhibitor. Inhibits NO production by activated macrophages. Selective Fabl bacterial enoyl-acyl carrier protein ACP reductase inhibitor. Inducer of G0/G1 cell cycle arrest and apoptosis in human A549 cells.
Catalog No. 50-256-1926
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.