Learn More
Cepharanthine, Thermo Scientific™
Specifications
Specifications
| Chemical Name or Material | Cepharanthine |
| CAS | 481-49-2 |
| Quantity | 1 g |
| Shipping Condition | No special requirements |
| Product Line | Life science |
| Molecular Formula | C37 H38 N2 O6 |
| Physical Form | Crystalline Powder |
| Color | White to Light-yellow to Beige |
| Purity | ≥94% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.