Learn More
Sigma Organic Chemistry Ceramic Etchant A | 500ML | MFCD08705343
Supplier: Sigma Organic Chemistry 667447500ML

Ceramic Etchant A, 500ML
About This Item:
MDL Number: MFCD08705343
UNSPSC Code: 12352300
Storage: room temp
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.