Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Ceramic Magnets
SDPSCI_ED

Catalog No. p-200009351 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Dimensions (L x W x H):
50 x 10 x 7 mm
70 x 15 x 8 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
S15906 50 x 10 x 7 mm
S15888 70 x 15 x 8 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S15906 Supplier United Scientific™ Supplier No. CBM050
Only null left
Add to Cart
Add to Cart

Simple strong magnets for the classroom.

  • We offer a variety of economical ceramic bar, disc, and ring shaped magnets.
  • Magnets are unpainted, except where noted.

Specifications

Material Ceramic
For Use With (Application) Magnetic Demonstrations
Grade 5+
Product Type Magnet
Dimensions (L x W x H) 50 x 10 x 7 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.