Learn More
QA Bio Inc Ludger Ceramide glycanase
Supplier:  QA Bio Inc LZCERHMKIT
Encompass
Ceramide glycanase is an enzyme used to release glycans from glycosphingolipids (GSLs) to enable their characterization. It cleaves glycans including GM1, GM2 and GM3 by cleaving the β-glycosyl linkage. Glycans can then be labelled using LudgerTag labeling technology. • Source: Hirudo medicinalis • Number of Samples: 25 samples • Amount of Samples: Up to 1 nmol of glycolipid • Suitable of Samples: Isolated glycolipids containing a ceramide backbone.
Catalog No. 50-223-6742
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.