Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ceric Sulfate, Reagent, 85%, Spectrum™ Chemical
SDP

Catalog No. 18601543 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
20 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-543 20 kg Fiber Drum
18-601-541 100 g Poly Bottle
18-601-542 500 g Poly Bottle
18-601-544 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-601-543 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C120420KGBL
Only null left
Add to Cart
Add to Cart
Request a Quote

C1204, 13590-82-4, Ce(SO4)2

Spectrum™ Chemical Ceric Sulfate, Reagent is an inorganic compound that is a yellow solid that is water soluble and used as a potent oxidizer. It is used for redox titration in analytical chemistry. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 13590-82-4
Molecular Formula CeO8S2
Molecular Weight (g/mol) 332.23
MDL Number MFCD00148852
InChI Key VZDYWEUILIUIDF-UHFFFAOYSA-J
IUPAC Name λ⁴-cerium(4+) disulfate
SMILES [Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

CAS Max % 100%
Quantity 20 kg
Percent Purity 85%
Grade Reagent
Insoluble Matter 0.1%
Packaging Fiber Drum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.