Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ 4-Fluoro-alpha-POP HCl (4-Fluoro PV9 HCl), 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: Intermetro Industries Corp 1842NFST316
Description
A novel synthetic cathinone derivative, 4-Fluoro-a-POP (4-Fluoro PV9) is an emerging drug of abuse that has been detected along side a-PVP in illegal drug products. Anecdotal reports suggest that 4-Fluoro-a-POP exerts similar stimulatory effects as a-PVP. Suitable uses for this Certified Spiking Solution™ include as starting material for calibrators and controls in synthetic cathinone testing methods by LC/MS or GC/MS for clinical toxicology, urine drug testing, or forensic analysis applications.Specifications
67-56-1 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
327.86 | |
1 mL |
C18H26FNO · HCl | |
1.0 mg/mL in methanol | |
Ampule |
Safety and Handling
Recommended Storage : -20°C
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation
Spot an opportunity for improvement?Share a Content Correction