Learn More
CERILLIANT™ (+/-)-Bromo-DragonFLY-D5 Hydrochloride Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier:  Intermetro Industries Corp SEC56K3SL
Catalog No. 01-001-046
Only null left
Add to Cart
Description
Description
Bromo-DragonFLY is an analog of the 2C amine family that also includes 2C-B, 2C-I, and 2C-E. This designer drug is an extremely potent hallucinogen with duration of action much longer than LSD. Toxicity of Bromo-DragonFLY appears to be fairly high for humans when taken in larger doses. This stable-labeled internal standard is suitable for quantitation of Bromo-DragonFLY levels in urine, serum, or plasma by GC/MS or LC/MS methods for use in clinical toxicology, forensic analysis, or urine drug testing applications.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
CERILLIANT is a registered trademark of Cerilliant Corporation